Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect ATM (pSer1981) Clone: 10H11.E12,

Catalog No. DR100250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
DR100250UG 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DR100250UG Supplier MilliporeSigma™ Supplier No. DR100250UG

Mouse Monoclonal Antibody

Specifically detects ATM (pSer1981) Clone: 10H11.E12 in Human, Mouse samples, and it is validated for Immunocytochemistry

ATM (ataxia telangiectasia mutated, ACC# NP_000042) is a member of the PI-3/PI-4 kinase family and is a central mediator of the cellular response to DNA damage and cellular stress. Mutations in this gene are associated with ataxia telangiectasia, an autosomal recessive disorder. ATM is a regulator of numerous downstream proteins, including the tumor suppressor proteins, p53 and BRCA1, the checkpoint kinase, CHK2, the checkpoint proteins RAD17 and RAD9, and the DNA repair protein NBS1. ATM and the functionally related ATR kinase are thought to be master regulators of cell cycle checkpoint signaling pathways that are required for the cellular response to DNA damage and as well as genome stability. Activation of ATM in response to DNA damage requires intermolecular autophosphorylation of Ser1981 and dimer dissociation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATM (pSer1981)
Applications Immunocytochemistry
Classification Monoclonal
Clone 10H11.E12
Conjugate Unconjugated
Formulation In PBS.
Host Species Mouse
Immunogen a synthetic phosphopeptide (SLAFEEGpSQSTTISS) corresponding to amino acids 1974-1988 surrounding the Ser1981 phosphorylation site of human ATM
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽370kDa ATM protein phosphorylated on Ser1981 in camptothecin-treated HeLa cells. Sold under license of U.S. Patent 7,108,992.
Target Species Human, Mouse
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.