Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect c-Raf (pTyr340, Tyr341), Polyclonal

Catalog No. 55300910T Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 Tests
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
55-300-910T 10 Tests Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-300-910T Supplier MilliporeSigma™ Supplier No. 55300910T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PhosphoDetect™ c-Raf (pTyr340, Tyr341) Clone: in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry

Theraf family of proto-oncogenes consists of three members: A-raf-1, B-raf, and c-raf-1 which show the greatest degree of homology in the carboxy-terminal half of the molecule. In the case of c-raf-1 this region has been shown to have protein (serine/threonine) kinase activity. C-raf was first identified in an oncogeneic form, v-raf, which is the oncogene of the transforming virus MSV. The raf-1 gene is the most studied of the three and encodes a protein with an apparent molecular mass of∽74kDa. Raf is widely expressed and plays an important role in cell proliferation and differentiation. Raf relays signals related to the activation of growth factor receptor to the activation of MAPK and utlimately to the nucleus. Activation of raf itself is dependent on tyrosine (341) phosphorylation. Tyrosine phosphorylation is accomplished by Src tyrosine kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PhosphoDetect™ c-Raf (pTyr340, Tyr341)
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation In phosphate buffer (without Mg2+ and Ca2+), 1mg/mL BSA, 50% glycerol, pH 7.3 (+/- 0.1).
Host Species Rabbit
Immunogen a synthetic phosphopeptide corresponding to amino acids surrounding the Tyr340 and Tyr341, phosphorylation sites of human c-Raf (Raf-1)
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes c-Raf phosphorylated at Tyr340 and Tyr341 in HEK293 cells overexpressing c-Raf.
Target Species Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.