Learn More
MilliporeSigma™ anti-PhosphoDetect Cdk1 (pTyr15), Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ 219440100UL
Description
Specifically detects PhosphoDetect™ Cdk1 (pTyr15) Clone: in Human, Rat samples, and it is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting
Entry of all eukaryotic cells into M-phase of the cell cycle is regulated by activation of Cdk1 kinase. Activation of Cdk1 is controlled at several steps including cyclin binding and phosphorylation of threonine161. However, the critical regulatory step in activating Cdk1 during progression into mitosis appears to be dephosphorylation of Tyr15 and Thr14. Phosphorylation at Tyr15 and inhibition of Cdk1 is carried out by WEE1 and MIK protein kinases while Tyr15 dephosphorylation and activation of Cdk1 is carried out by the Cdc25 phosphatase.
Specifications
PhosphoDetect™ Cdk1 (pTyr15) | |
Polyclonal | |
In 150mM NaCl, 10mM sodium HEPES, 100mg/mL BSA, 50% glycerol, pH 7.5. | |
a synthetic phosphopeptide corresponding to amino acids surrounding the Tyr15 phosphorylation site of human Cdk1 | |
RUO | |
Recognizes the ∽34kDa Cdk1 protein, phosphorylated at Tyr15, in hydroxyurea treated SaoS cell extracts. Also recognizes phosphorylated Cdk2. Does not react with Cdk4, Cdk6, Cdk7, or other phosphotyrosine proteins. | |
Purified |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
100 μL | |
Primary | |
Human, Rat | |