Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect EGFR (pTyr1068), Polyclonal

Catalog No. 32486710T Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 Tests
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
32-486-710T 10 Tests Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-486-710T Supplier MilliporeSigma™ Supplier No. 32486710T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PhosphoDetect™ EGFR (pTyr1068) Clone: in Chicken, Human, Mouse, Rat samples, and it is validated for Immuno Blotting

Epidermal Growth Factor (EGF) is responsible for a variety of biological effects including mitogenesis and differentiation. The EGF receptor (EGFR) has three domains: the extracellular, transmembrane, and cytoplasmic domains. Upon binding of EGF to the extracellular domain, dimerization of the receptor occurs and the cytoplasmic domains display tyrosine kinnase activity. This leads to autophosphorylation of the∽170kDa glycoprotein on various residues within the cytoplasmic domain as well as phosphorylation of intracellular substrates. Tyr1068 is an autophosphorylation site in EGFR. The SH2 domain of Grb2 binds to Tyr1068 of EGFR in the multimeric complex EGFR-Grb2-mSos1 that is formed upon stimulation of EGFR with EGF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PhosphoDetect™ EGFR (pTyr1068)
Applications Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation In Dulbecco's PBS, 1mg/mL BSA, 50% glycerol, pH 7.4.
Host Species Rabbit
Immunogen a synthetic phosphopeptide corresponding to amino acids surrounding the Tyr1068 phosphorylation site of human EGFR
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽185kDa EGFR protein phosphorylated at Tyr1068. Note: 1 T = 1 test. Sufficient for 10 Western miniblots.
Target Species Chicken, Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.