Learn More
MilliporeSigma™ anti-PhosphoDetect EGFR (pTyr1173) Clone: 9H2,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ 3248641SET
Description
Specifically detects EGFR (pTyr1173) Clone: 9H2 in Human, Mouse, Rat samples, and it is validated for ELISA, Immunoprecipitation, Immuno Blotting
Binding of EGF to the extracellular domain of the EGF-receptor results in receptor dimerization and autophosphorylation on tyrosine residues. The major site of tyrosine phosphorylation is tyrosine 1173.
Specifications
EGFR (pTyr1173) | |
Monoclonal | |
Unconjugated | |
Mouse | |
1 Set | |
Primary | |
Human, Mouse, Rat | |
Purified |
ELISA, Immunoblot, Immunoblot, Immunoprecipitation | |
9H2 | |
100μg antibody lyophilized from 2X PBS, PEG, sucrose and 200μL lyophilized control lysate. | |
a synthetic phosphopeptide (NAEpYLRV) corresponding to amino acids 1170-1176 of human EGFR | |
RUO | |
Recognizes EGFR phosphorylated at Tyr1173 in vanadate-treated HepG2 cells. | |
−20°C | |
IgG1 |