Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect FAK (pTyr397), Polyclonal

Catalog No. 34129210T Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 Tests
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
34-129-210T 10 Tests Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-129-210T Supplier MilliporeSigma™ Supplier No. 34129210T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PhosphoDetect™ FAK (pTyr397) Clone: in Chicken, Drosophila, Frog, Human, Mouse, Rat samples, and it is validated for Immunohistochemistry

Discovered as a substrate for Src, FAK acts as a key element of integrin signaling. FAK is a non-receptor tyrosine kinase that plays a central role in cell spreading, differentiation, migration, cell death, and acceleration of the G1 to S phase transition of the cell cycle. The Tyr397 autophosphorylation site binds Src family SH2 and the p85 subunit of PI3-Kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PhosphoDetect™ FAK (pTyr397)
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation In Dulbecco's PBS (no Mg2+ and Ca2+), 1mg/mL BSA, 50% glycerol, pH 7.3.
Host Species Rabbit
Immunogen a synthetic phosphopeptide corresponding to amino acids surrounding the Tyr397 phosphorylation site of human FAK
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽125kDa FAK protein when phosphorylated at Tyr397. Note: 1 T = 1 test. Sufficient for 10 Western miniblots.
Target Species Chicken, Drosophila, Frog, Human, Mouse, Rat
Content And Storage ≤ -70°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.