Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect GSK-3alpha/beta (pTyr279/216), Polyclonal

Catalog No. ST101310T Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 Tests
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST101310T 10 Tests Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST101310T Supplier MilliporeSigma™ Supplier No. ST101310T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PhosphoDetect™ GSK-3α/β (pTyr279/216) Clone: in Human, Mouse, Rat samples, and it is validated for Dot Blot, ELISA, Immunocytochemistry, Immuno Blotting

GSK-3α contains a phosphate on tyrosine 279, and GSK-3β contains a phosphate on tyrosine 216. GSK is a protein serine kinase involved in the control of regulatory proteins like glycogen synthase and the transcription factor c-Jun. It has also been associated with the regulation of the microtubule-binding protein Tau, and may be involved in the pathogenesis of Alzheimer's Disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PhosphoDetect™ GSK-3α/β (pTyr279/216)
Applications Dot Blot, ELISA, Immunoblot, Immunoblot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation In Dulbecco's PBS (without Mg2+ and Ca2+), 50% glycerol, 1mg/mL BSA, pH 7.3.
Host Species Rabbit
Immunogen a synthetic phosphopeptide corresponding to amino acids surrounding the Tyr279/216 phosphorylation sites of human GSK-3α/β
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽51kDa GSK-3α protein phosphorylated at Tyr279 and the ∽47kDa protein GSK-3β phosphorylated at Tyr216.
Target Species Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.