Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect GSK3beta (pSer9) Clone: 2D3,

Catalog No. 3615271SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
36-152-71SET 1 Set Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-152-71SET Supplier MilliporeSigma™ Supplier No. 3615271SET
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects GSK3β (pSer9) Clone: 2D3 in Human, Mouse samples, and it is validated for ELISA, Immuno Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen GSK3β (pSer9)
Applications ELISA, Immunoblot, Immunoblot
Classification Monoclonal
Clone 2D3
Conjugate Unconjugated
Formulation 100μg antibody in PBS, PEG, sucrose, 50% glycerol and 200μL lyophilized control lysate.
Host Species Mouse
Immunogen a synthetic phosphopeptide corresponding to amino acids surrounding the Ser9 phosphorylation site of human glycogen synthase kinase 3β
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽46kDa GSK-3β phosphorylated at Ser9. Supplied with a control lysate derived from 2 x 106 EGF-treated HepG2 cells.
Target Species Human, Mouse
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.