Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect Histone H3 (pSer10) (7-20), Polyclonal

Catalog No. 38215950UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
38-215-950UG 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-215-950UG Supplier MilliporeSigma™ Supplier No. 38215950UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PhosphoDetect™ Histone H3 (pSer10) (7-20) Clone: in C. elegans, Drosophila, Human, Tetrahymena, Xenopus samples, and it is validated for Immunocytochemistry, Immunohistochemistry, Immunoprecipitation

Histone H1 and H3 phosphorylations correlate with the process of chromosome condensation. H3 mRNA-positive cells are most often found in tissues with a high proliferation rate, such as basal cells of the skin and crypt cells of the intestine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PhosphoDetect™ Histone H3 (pSer10) (7-20)
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation In 150mM NaCl, 100mM Tris-glycine, pH 7.4.
Host Species Rabbit
Immunogen a synthetic phosphopeptide [ARKpSTGGKAPRKQL(C)] corresponding to amino acids 7-20 surrounding the Ser10 phosphorylation site of human histone H3, conjugated to KLH
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽17kDa histone H3 protein phosphorylated at Ser10.
Target Species C. elegans, Drosophila, Human, Tetrahymena species, Xenopus species
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.