Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect NMDA NR2B (pTyr1472), Polyclonal

Catalog No. 454583100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
45-458-3100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-458-3100UL Supplier MilliporeSigma™ Supplier No. 454583100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PhosphoDetect™ NMDA NR2B (pTyr1472) Clone: in Rat samples, and it is validated for Dot Blot, Immuno Blotting

NMDA NR2B is phosphorylated at Tyr1472 by Src-family tyrosine kinases such as Fyn and may be important for synaptic plasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PhosphoDetect™ NMDA NR2B (pTyr1472)
Applications Dot Blot, Immunoblot, Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 150mM NaCl, 10mM HEPES, 100μg/mL BSA, 50% glycerol, pH 7.5.
Host Species Rabbit
Immunogen a synthetic phosphopeptide corresponding to amino acids surrounding the Tyr1472 phosphorylation site of rat NMDA NR2B
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽180kDa NMDA receptor NR2B subunit phosphorylated at Tyr1472.
Target Species Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.