Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-PhosphoDetect PDH-E1alpha (pSer300), Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ AP106450UG
Description
Specifically detects PhosphoDetect™ PDH-E1α (pSer300) Clone: in Human samples, and it is validated for Immunocytochemistry
The pyruvate dehydogenase (PDH) complex (PDC) catalyzes the irreversible conversion of pyruvate to acetyl CoA, providing the link between glycolyis and the TCA cycle. Dysregulation of the PDC in skeletal muscle has been implicated in type 2 diabetes and mutations in the PDC result in various mitochondrial diseases. The PDC is a multienzyme complex composed of several copies of three catalytic proteins (E1, E2, and E3): one structural (E3-binding protein) and two regulatory proteins (PDH kinase [PDK] and PDH phosphatase [PDP]). The activity of the PDC is mainly regulated by phosphorylation-dephosphorylation of three specific serine residues (232, 293, 300) on the E1α subunit. Phosphorylation of PDH determines the activity of the complex. Phosphorylation by one of four PDK isoforms inactivates the complex, while removal of the phosphate by one of two PDP isoforms activates the complex. The activity of the PDP1 isoform is sensitive to Ca2+, while the PDK isoforms are regulated by the reduction and acetylation status of the E2 subunit, pyruvate, and the small molecule the anti cancer dichloroacetate. (DCA).
Specifications
PhosphoDetect™ PDH-E1α (pSer300) | |
Polyclonal | |
In 150mM NaCl, 100mM Tris-Glycine, pH 7.4. | |
a synthetic phosphopeptide corresponding to amino acids surrounding the Ser300 phosphorylation site of human PDH-E1α | |
RUO | |
Recognizes the ∽44kDa PDH-E1α protein phosphorylated at Ser300 in HEK293 cells. | |
−20°C |
Immunocytochemistry | |
Unconjugated | |
Rabbit | |
50 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction