Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect Phosphotyrosine Clone: PY20,

Catalog No. 525295100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
52-529-5100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-529-5100UG Supplier MilliporeSigma™ Supplier No. 525295100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Phosphotyrosine Clone: PY20 in Multi samples, and it is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Immuno Blotting

Cellular activation, proliferation and differentiation are signaled by phosphorylation of tyrosine residues. Growth factor receptors and oncogene protein kinases are associated with tyrosine phosphorylation. Anti-phosphotyrosine antibodies are indispensable for the study of cellular tyrosine phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphotyrosine
Applications ELISA, Immunoblot, Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone PY20
Conjugate Unconjugated
Formulation In 10mM sodium phosphate, pH 7.4.
Host Species Mouse
Immunogen KLH conjugated to phosphotyrosine, glycine, and alanine
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes a broad spectrum of tyrosine-phosphorylated proteins in their native and denatured states in crude extracts.
Target Species Multi-species
Content And Storage −20°C
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.