Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect PKR (pThr451), Polyclonal

Catalog No. 52746010T Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 Tests
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
52-746-010T 10 Tests Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-746-010T Supplier MilliporeSigma™ Supplier No. 52746010T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PhosphoDetect™ PKR (pThr451) Clone: in Human, Mouse samples, and it is validated for Immunohistochemistry

PKR is an interferon-inducible serine/threonine kinase activated by double stranded RNA. The activation of PKR is accompanied by phosphorylation at multiple sites, including Thr446 and Thr451 in the PKR activation loop. PKR phosphorylates theα-subunit of the eukaryotic initiation factor-2 and inhibits protein synthesis. PKR also phosphorylates p53 and plays a role in the interferon response to viral infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PhosphoDetect™ PKR (pThr451)
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation In Dulbecco's PBS (without Mg+2 or Ca+2), 0.1% BSA, 50% glycerol, pH 7.3.
Host Species Rabbit
Immunogen a synthetic phosphopeptide corresponding to amino acids surrounding the Thr451 phosphorylation site of human PKR (double-stranded RNA-dependent protein kinase)
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽65-68kDa PKR protein phosphorylated at Thr451.
Target Species Human, Mouse
Content And Storage ≤ -70°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.