Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect SAPK/JNK (pThr183, Tyr185), Polyclonal

Catalog No. 55930910T Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 Tests
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
55-930-910T 10 Tests Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-930-910T Supplier MilliporeSigma™ Supplier No. 55930910T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PhosphoDetect™ SAPK/JNK (pThr183, Tyr185) Clone: in C. elegans, Chicken, Drosophila, Human, Mouse, Rat samples, and it is validated for Immunohistochemistry

C-Jun-N-terminal kinase (JNK), also called stress-activated protein kinase (SAPK) is a member of the mitogen activated protein kinase (MAPK) family. JNK is activated in response to various extracellular stimuli, including pro-inflammatory cytokines. Phosphorylation of threonine and tyrosine in the Thr-Pro-Tyr domain is required for full activation. Mitogen activated protein kinase kinase (MKK4) preferentially phosphorylates tyrosine 185, and MKK7 causes preferential phosphorylation of threonine 183.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PhosphoDetect™ SAPK/JNK (pThr183, Tyr185)
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation In Dulbecco's PBS (without Mg2+ and Ca2+), 50% glycerol, 1mg/mL BSA, pH 7.3.
Host Species Rabbit
Immunogen a synthetic phosphopeptide corresponding to amino acids surrounding the Thr183 and Tyr185 phosphorylation sites of human SAPK/JNK
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽49kDa SAPK1 protein and the ∽55kDa SAPK2 protein phosphorylated at Thr183 and Tyr185. Antibody Target Gene Symbol: MAPK8, MAPK9 Target Synonym: AI849689, C-JUN N-TERMINAL KINASE1, JNK, JNK1, JNK1 PROTEIN KINASE, JNK1A2, JNK21B1/2, p46JNK1, p46JNK1 ALPHA, PRKM8, Sapk gamma, SAPK P46, SAPK1, SAPK1/JNK, STRESS-ACTIVATED PROTEIN KINASE-LIKE KINASE, AI851083, Jnk p54, Jnk p55, JNK-55, JNK2, JNK2A, Jnk2a2, JNK2ALPHA, JNK2B, JNK2BETA, JUNKINASE, p54a, p54aSAPK, PRKM9, SAPK, SAPK ALPHA, SAPK ALPHA 1 Entrez Gene Name: mitogen-activated protein kinase 8
Target Species C. elegans, Chicken, Drosophila, Human, Mouse, Rat
Content And Storage ≤ -70°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.