Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect Shc/p66 (pSer36) Clone: 60000000000,

Catalog No. 5668071SET Shop All MilliporeSigma Products
missing translation for 'changeView'
Click to view available options
Quantity:
1 Set
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
56-680-71SET 1 Set Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 56-680-71SET Supplier MilliporeSigma™ missing translation for 'supplierNo' 5668071SET
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Shc/p66 (pSer36) Clone: 60000000000 in Human, Mouse samples, and it is validated for ELISA, Immuno Blotting

Mammalian cells can express three alternatively spliced isoforms of the shc adaptor protein: Shc/p46, Shc/p52 and Shc/p66. Shc/p46 and Shc/p52 are found in every cell type, whereas Shc/p66 expression varies in different tissues. Shc proteins are phosphorylated by receptor tyrosine kinases and interact with phosphotyrosine residues on these receptors via the shc PTB and/or SH2 domains. The major tyrosine phosphorylation sites are tyrosines 239/240 as well as tyrosine 317. The Shc/p66 protein contains an additional N-terminal protein domain that is phosphorylated at serine 36. It is suggested that only the serine-phosphorylated Shc/p66 can interact with GRB2. Moreover, expression of Shc/p66 accelerates the inactivation of MAP kinases after EGF stimulation, whereas Shc/p46 and Shc/p52 enhance MAP kinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Shc/p66 (pSer36)
Applications ELISA, Immunoblot, Immunoblot
Classification Monoclonal
Clone 60000000000
Conjugate Unconjugated
Formulation 100μg antibody lyophilized from 2X PBS, PEG, sucrose and 125μL control protein lyophilized from 2X PBS.
Host Species Mouse
Immunogen a synthetic phosphopeptide corresponding to amino acids surrounding the Ser36 phosphorylation site of human Shc/p66
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽66kDa form of Shc phosphorylated at Ser36. Does not cross-react with the non-phosphorylated form of Shc/p66 or with unrelated phosphorylation sites.
Target Species Human, Mouse
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.