Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect STAT6 (pTyr641), Polyclonal

Catalog No. 575143100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
57-514-3100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-514-3100UL Supplier MilliporeSigma™ Supplier No. 575143100UL

Rabbit Polyclonal Antibody

Specifically detects PhosphoDetect™ STAT6 (pTyr641) Clone: in Human, Mouse samples, and it is validated for Flow Cytometry, Immunofluorescence, Immuno Blotting

STAT (signal transducer and activation of transcription) factors are central components of signaling cascades triggered by cytokine receptors. STAT6 is activated in response to IL-4, and it plays a central role in exerting IL-4 mediated biological responses. STAT6 is phosphorylated on Tyr641 in response to ligand stimulation via receptor-associated Janus Kinases (JAKs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PhosphoDetect™ STAT6 (pTyr641)
Applications Flow Cytometry, Immunoblot, Immunoblot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation In 150mM NaCl, 10mM HEPES, 100μg/mL BSA, 50% glycerol, pH 7.5.
Host Species Rabbit
Immunogen a synthetic phosphopeptide corresponding to amino acids surrounding the Tyr641 phosphorylation site of human STAT6, conjugated to KLH
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽94kDa STAT6 protein phosphorylated at Tyr641.
Target Species Human, Mouse
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.