Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-PhosphoDetect Tie2 (pTyr 1108/1117) (Ab-2), Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ PC45025UL
Description
Specifically detects PhosphoDetect™ Tie2 (pTyr,1108/1117) Clone: in Human samples, and it is validated for ELISA, Immunohistochemistry (Paraffin)
Angiopoietin-1 (Ang1) and its naturally occurring inhibitor, angiopoietin-2 (Ang2) exert their effects by inducing autophosphorylation of their signaling receptor Tie2 (also known as Tek). Phosphorylation of the receptor occurs during vasculogenesis and is required for its activity in regulating endothelial cell proliferation, differentiation and guiding the proper patterning of endothelial cells during blood vessel formation. Tie-2 is a∽140kDa transmembrane receptor-tyrosine kinase that is expressed by endothelial cells and primitive hematopoietic tissues. Tie2/Tek has been shown to be critical for endothelial cell survival, which is evidenced in part by the constitutive phosphorylation that is observed in quiescent endothelium. More recently a multifunction docking site has been identified on the receptor (Tyr1100in mouse or Tyr1102in human) that serves as a binding site for other partners including Grb2, Grb7, and p85 and acting via an SH2 domain interaction.
Specifications
PhosphoDetect™ Tie2 (pTyr,1108/1117) | |
Polyclonal | |
Undiluted serum. | |
a synthetic phosphopeptide [pYEKFTpYAGID(C)] corresponding to amino acids surrounding the Tyr1108/1117 phosphorylation sites 1108-1117 of human Tie2 | |
RUO | |
Recognizes the ∽140-145 Tie2 protein phosphorylated at Tyr1108 and Tyr1117 in colon carcinoma tissue. | |
−20°C | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
25 μL | |
Primary | |
Human | |
Undiluted |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction