Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect VASP (pSer157) Clone: 5C6,

Catalog No. 676604100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
67-660-4100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-660-4100UG Supplier MilliporeSigma™ Supplier No. 676604100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects VASP (pSer157) Clone: 5C6 in Human, Mouse samples, and it is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Immuno Blotting

VASP (vasodilator-stimulated phosphoprotein), an established substrate of cAMP- and cGMP-dependent protein kinases in vitro and in living cells, is associated with focal adhesions, microfilaments, and membrane regions of high dynamic activity. VASP binds in vitro to profilin, zyxin, vinculin, and the Listeria sp. surface protein ActA. VASP also promotes actin nucleation and binds to actin filaments in vitro and associates with stress fibers in cells. Studies imply that VASP phosphorylation may act as an immediate negative regulator of actin dynamics. Phosphorylation of Ser157 in VASP can be used as a measure of PKA activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VASP (pSer157)
Applications ELISA, Immunoblot, Immunoblot, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 5C6
Conjugate Unconjugated
Formulation Lyophilized from PBS, PEG, sucrose.
Host Species Mouse
Immunogen a synthetic phosphopeptide corresponding to amino acids surrounding the Ser157 phosphorylation site of human VASP
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the the ∽50kDa VASP protein phosphorylated at Ser157.
Target Species Human, Mouse
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.