Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PhosphoDetect VEGFR2 (pTyr1054/1059), Polyclonal

Catalog No. PS101310T Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 Tests
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
PS101310T 10 Tests Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PS101310T Supplier MilliporeSigma™ Supplier No. PS101310T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PhosphoDetect™ VEGFR2 (pTyr1054/1059) Clone: in Mouse samples, and it is validated for Immuno Blotting

Vascular Endothelial Growth Factor Receptor 2 (VEGFR2) is receptor tyrosine kinase that serves as a key mediator in angiogenesis and plays an essential role in embryonic development, wound healing, cell proliferation, migration, and differentiation. It contains seven immunoglobulin-like sequences in the extracellular domain and exhibits sequence similarity to Flt-1 (FMS-like tyrosine kinase 1) and Flt-4 (FMS-like tyrosine kinase 4). Upon binding to its ligand, VEGF, VEGFR2 undergoes ligand-induced dimerization, which activates its intrinsic tyrosine kinase activity resulting in auto- and trans-phosphorylation on specific tyrosine residues in the cytoplasmic domain. These tyrosine residues, when phosphorylated, are involved as docking sites to recruit molecules containing Src homology 2 (SH2) or phosphotyrosine-binding domains and to convey signals to downstream effectors. Phosphorylation of VEGFR2 at Tyr1054 and Tyr1059 within the activation loop is required for required for activation of VEGFR2 and enhances its intrinsic tyrosine kinase activity. Phosphorylation at Tyr1059 is also responsible for VEGF-induced intracellular Ca2+ mobilization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PhosphoDetect™ VEGFR2 (pTyr1054/1059)
Applications Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation In Dulbecco's PBS (without Mg2+ and Ca2+), 1mg/mL BSA, 50% glycerol, pH 7.3 (± 0.1).
Host Species Rabbit
Immunogen a synthetic phosphopeptide corresponding to amino acids surrounding the Tyr1054/1059 phosphorylation sites of mouse and human VEGFR-2
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽200kDa VEGFR-2 protein phosphorylated at Tyr1054/1059 in porcine pancreatic cells transfected with a CSF-1/mouse VEGFR-2 chimeric receptor.
Target Species Mouse
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.