Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Phospholamban, Clone: A1, ™ (Upstate™)
Supplier: MilliporeSigma™ 05205/DEL

Description
Suzuki et al., Biochem. Cell Biol. 65:302, 1987
ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications
Phospholamban | |
Unconjugated | |
Bovine cardiac phospholamban | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
50 μg | |
Endocrine/Metabolism | |
Bovine, Canine, Mouse, Pig, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction