Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-lipase A2 Mouse anti-Human, Mouse, Rat, Clone: CH-7, MilliporeSigma™

Catalog No. 051406MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-140-6MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-140-6MI Supplier MilliporeSigma Supplier No. 051406
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Phospholipase A2 Antibody, clone CH-7 detects level of Phospholipase A2 and has been published and validated for use in western blot, ELISA, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospholipase A2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CH-7
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2b in buffer containing 20mM sodium phosphate, 250mM sodium chloride, 0.1% sodium azide, pH. 7.6.
Gene Accession No. NM_000928.2
Gene Symbols PLA2G1B, MGC119834, PPLA2, PLA2, PLA2A, MGC119835, EC 3.1.1.4
Host Species Mouse
Immunogen Recombinant cPLA2
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.