Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-lipase A2 Mouse anti-Recombinant Human Protein, Clone: CH-7, MilliporeSigma™

Catalog No. MAB1668MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1668MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1668MI Supplier MilliporeSigma Supplier No. MAB1668
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Phospholipase A2 using this Anti-Phospholipase A2 Antibody, clone CH-7 validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospholipase A2
Applications ELISA, Western Blot
Classification Monoclonal
Clone CH-7
Conjugate Unconjugated
Formulation In 20mM sodium phosphate, 250mM sodium chloride, 0.1% sodium azide, pH. 7.6
Gene Accession No. NM_000928.2
Gene Symbols PLA2G1B, MGC119834, PPLA2, PLA2, PLA2A, MGC119835, EC 3.1.1.4
Host Species Mouse
Immunogen Recombinant cPLA2 expressed from the human cPLA2 gene.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.