Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospholipase Cγ1 Mouse, Unlabeled, Clone: 10/PLCGAMMA, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610027
Description
The Phospholipase C (PLC) isozymes hydrolyze phosphatidyl inositol biphosphate to inositol triphosphate and diacylglycerol. The former causes release of calcium from the endoplasmic reticulum, while the latter is an activator of Protein Kinase C. Within the PLC family, PLCγ is the only member that contains SH2 and SH3 domains. These domains enable it to interact with receptor tyrosine kinases and become enzymatically activated via phosphorylation. It exists as two isoforms: 1) PLCγ 1, which is ubiquitously expressed, and 2) PLCγ 2, found primarily in the lymphoid system. PLCγ is essential for growth factor-induced cell motility and mitogenesis. PLCγ 1 null mice exhibit retarded embryonic growth and lethality in midgestation. Overexpression of PLCγ is evident in several forms of cancer, and it has been identified as a key mediator of PDGF-dependent cellular transformation. Thus regulation of PLCγ activity by growth factors is involved in cell growth and transformation.
Specifications
| Phospholipase Cγ1 | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Cow PLCγ1 N-terminal region Peptide | |
| 50 μg | |
| Cell Biology | |
| Canine, Chicken, Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| 10/PLCGAMMA | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction