Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospholipase Cβ1 Mouse, Unlabeled, Clone: 16, BD

Catalog No. BDB610924 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610924 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610924 Supplier BD Biosciences Supplier No. 610924

Mouse Monoclonal Antibody

Phospholipase C (PLC) hydrolyzes inositol phospholipids into diacylglycerol (DAG) and inositol 1,4,5-trisphosphate (IP3). The former causes release of Ca2+ from intracellular stores, while the latter is an activator of PKC. At least three isozymes are known: PLCβ, PLCγ, and PLCδ. They recognize phosphatidylinositol (PI), phosphatidylinositol 4-phosphate (PIP2), and phospatidylinositol 4,5 bisphosphate (PIP2) and carry out the Ca2+-dependent hydrolysis of these inositol phospholipids. All three classes contain two highly conserved regions, designated “X” and “Y”, whose structural integrity is essential for a functional catalytic core. The PLCβ subfamily has been reported to contain at least four members: β1, β2, β3 and β4. PLCβ1 has been reported to have been identified and cloned from brain tissue where the PLCβ1 gene encodes for two different mRNAs. The resulting protein products, PLCβ1a (121 aa) and PLCβ1b (1173 aa) differ only at their C-termini. Although PLCγ activity is induced by receptor tyrosine kinase phosphorylation, PLCβ is regulated by G protein α subunits or βγ subunits. In particular, Gαq has been reported to activate PLCβ1.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospholipase Cβ1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 16
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Rat Phospholipase Cβ1 aa. 4-159
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.