Learn More
Phosphoserine/threonine Mouse, Unlabeled, Clone: 22A, BD
Mouse Monoclonal Antibody
$400.00 - $675.00
Specifications
Antigen | Phosphoserine/threonine |
---|---|
Clone | 22A |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Protein phosphorylation of serine and threonine residues is critical for the control of protein activity involved in various cellular events. An assortment of Ser/Thr kinases and phosphatases regulate serine and threonine phosphorylation in cell signaling pathways, such as growth factor, cytokine, chemokine, and stress response. Detection of serine and threonine phosphorylation can generally be monitored by antibodies that detect phosphoserine and phosphothreonine.
Immunofluorescence, Western Blotting
Specifications
Phosphoserine/threonine | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
22A | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Rat | |
IgG1 | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.