Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-threonine-Proline Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 071473MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-147-3MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-147-3MI Supplier MilliporeSigma Supplier No. 071473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Phosphothreonine-Proline using this Anti-Phosphothreonine-Proline Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphothreonine-Proline
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in TBS with 0.05% sodium azide and 50% glycerol.
Host Species Rabbit
Immunogen KLH-phosphothreonine-proline amide (pT-P-NH2) peptide conjugates
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.