Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-tyrosine Mouse anti-Vertebrates, Clone: 4G10™, MilliporeSigma™

Catalog No. 16101MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-101-MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-101-MI Supplier MilliporeSigma Supplier No. 16101
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Phosphotyrosine Antibody, clone 4G10™, agarose conjugate is an antibody against Phosphotyrosine for use in immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphotyrosine
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Agarose
Formulation 1 mg of Anti-Phosphotyrosine monoclonal antibody covalently coupled to 1mL of protein A agarose beads and provided as a 50% slurry suspended in PBS, containing 0.05% ProClin™ 150 as a preservative, for a total volume of 2 mL. Liquid susp
Host Species Mouse
Immunogen Phosphotyramine-KLH.
Purification Method Protein A purfied
Quantity 1 mg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.