Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4G10™ Platinum, Phosphotyrosine Mouse anti-All, Biotin, Clone: 4G10™ , MilliporeSigma™

Catalog No. 16452MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-452-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-452-MI Supplier MilliporeSigma Supplier No. 16452
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

4G10 Platinum, Anti-Phosphotyrosine, Biotin Conjugate detects levels of Phosphotyrosine proteins and has been published and validated for use in immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphotyrosine
Applications Immunoprecipitation
Classification Monoclonal
Clone 4G10™
Conjugate Biotin
Formulation 100μL of a proprietary mixture of biotin conjugated, protein G purified mouse monoclonals 4G10™ (IgG2bκ) and PY20 (IgG2b) in PBS, pH 7.4 with 0.05% sodium azide.
Host Species Mouse
Immunogen The immunogen for 4G10™ was phospho-tyramine coupled to KLH, while the immunogen for PY20 was phosphotyrosine conjugated to a carrier protein.
Purification Method Protein G-Sepharose™ chromatography
Quantity 100 μL
Primary or Secondary Primary
Target Species All species
Content And Storage 2-8°C for 6 months. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.