Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-tyrosine Mouse anti-Vertebrates, Clone: 4G10™, MilliporeSigma™

Catalog No. 05777MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-777-MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-777-MI Supplier MilliporeSigma Supplier No. 05777
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Phosphotyrosine Antibody, clone 4G10 is a recombinant antibody that detects tyrosine phosphorylated proteins in all species. This unique recombinant antibody is validated for use in immunocytochemistry, immunohistochemistry, immunoprecipitation, western blotting and is published in peer review journals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphotyrosine
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4G10™
Conjugate Unconjugated
Formulation Protein G purified recombinant 4G10 mouse IgG2bk in 0.01M Phosphate Buffer containing no preservative, pH 7.1
Host Species Mouse
Immunogen Produced from CHO cells expressing the 4G10 antibody heavy and light chain cDNAs. Heavy chain C-terminus has a hexa-histidine tag for purification and immobilization via Nickel affinity matrices.
Purification Method Protein G purfied
Quantity 1 mg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.