Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-tyrosine Mouse anti-All, Clone: 4G10™, MilliporeSigma™

Catalog No. 16282MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-282-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-282-MI Supplier MilliporeSigma Supplier No. 16282
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Phosphotyrosine using this Anti-Phosphotyrosine Antibody, clone 4G10™, Magnetic Bead Conjugate validated for use in immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphotyrosine
Applications Immunoprecipitation
Classification Monoclonal
Clone 4G10™
Conjugate Unconjugated
Formulation 20% slurry; 1xPBS, 0.05% sodium azide. Sufficient for 10 IPs.
Host Species Mouse
Immunogen Phosphotyramine-KLH
Purification Method Protein G-Sepharose Chromatography
Quantity 200 μL
Primary or Secondary Primary
Target Species All species
Content And Storage 2-8°C for 12 months. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.