Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-tyrosine Mouse anti-All, Clone: 4G10™, MilliporeSigma™

Catalog No. 05321MGMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-321-MGMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-321-MGMI Supplier MilliporeSigma Supplier No. 05321MG
Add to Cart
Add to Cart
Request a Quote

Mouse Monoclonal Antibody

Anti-Phosphotyrosine Antibody, clone 4G10 detects level of Phosphotyrosine and has been published and validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphotyrosine
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4G10™
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing PBS, pH 7.4. Liquid at 2-8ºC.
IgG2bκ mouse monoclonal antibody produced in vitro by mouse-mouse hybridoma 4G10™ (FOX-NY [NS-1 derivative] myeloma x spleen cells).
Host Species Mouse
Immunogen Phosphotyramine-KLH
Purification Method Protein G-Sepharose™ chromatography
Quantity 1 mg
Primary or Secondary Primary
Target Species All species
Content And Storage 2-8°C for 12 months. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.