Learn More
MilliporeSigma™ anti-Phosphotyrosine, Clone: 4G10™, ™
Supplier: MilliporeSigma™ 16452/DEL
Description
The development of the anti-phosphotyrosine, clone 4G10™ in 1989 was a monumental discovery for researchers. 4G10™ was the first and best single monoclonal antibody for the detection tyrosine phosphorylation. 4G10™ is well known for its sensitivity and its ability to detect multiple tyrosine phosphorylations on numerous substrates. It has been validated by thousands of scientific and medical researchers in virtually every application and tyrosine target over the past 2 decades. Now Millipore has made the best even better with 4G10™ Platinum. We pooled 4G10™ with the next most highly regarded anti-phosphotyrosine, clone PY20, to make 4G10™ Platinum. PY20 itself is a very poor substitute for 4G10™, but its additive effect allow for a greater level of detection on more substrates that even 4G10™ alone is capable.
Host Species: Mouse
Clone: 4G10™
Isotype: IgG2bκ
Species Reactivity: All
Epitope: Phosphotyrosine
Immunogen: The immunogen for 4G10™ was phospho-tyramine coupled to KLH, while the immunogen for PY20 was phosphotyrosine conjugated to a carrier protein.
Formula Weight [Chemical]: Varies
Western Blotting
Specifications
| Phosphotyrosine | |
| 4G10™ | |
| Mouse | |
| 100 μL | |
| Cell Signaling | |
| Semi-purified |
| Monoclonal | |
| Biotin | |
| The immunogen for 4G10™ was phospho-tyramine coupled to KLH, while the immunogen for PY20 was phosphotyrosine conjugated to a carrier protein. | |
| RUO | |
| Primary | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.