Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Phosphotyrosine, Clone: 4G10 Platinum, ™ (Upstate™)
Specifications
Classification | Monoclonal |
---|---|
Conjugate | Unconjugated |
Form | Semi-purified |
Host Species | Mouse |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-171-464
|
MilliporeSigma™
051050X/DEL |
30 μL | N/A | N/A | N/A | ||||
50-171-463
|
MilliporeSigma™
051050/DEL |
100 μL | N/A | N/A | N/A | ||||
Description
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Immunoprecipitation Kinase Assay, Western Blotting
Specifications
Monoclonal | |
Semi-purified | |
RUO | |
IgG2b | |
Protein G-Sepharose chromatography | |
Yes |
Unconjugated | |
Mouse | |
Cell Signaling | |
Primary | |
IgG2b |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title