Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-tyrosine Mouse anti-Vertebrates, Clone: PY20, MilliporeSigma™

Catalog No. 05947MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-947-MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-947-MI Supplier MilliporeSigma Supplier No. 05947
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Phosphotyrosine Antibody, clone PY20 is an antibody against Phosphotyrosine for use in ELISA, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphotyrosine
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PY20
Conjugate Unconjugated
Formulation 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4. Liquid at 2-8°C.
Host Species Mouse
Immunogen Phosphotyrosine conjugated to carrier protein.
Purification Method Protein A purfied
Quantity 1 mg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.