Learn More
MilliporeSigma™ anti-Phosphotyrosine, Clone: PY20, HRP
Supplier: MilliporeSigma™ 16454/DEL

Description
Protein tyrosine phosphorylation was first described in 1979. Large numbers of tyrosine kinases have been described since, and many of the signaling pathways initiated by tyrosine phosphorylation have been delineated. Availability of a top-quality anti-phosphotyrosine antibody has allowed the study of tyrosine kinase activity by simple blotting procedures, and without routine need of large quantities of radioactivity.
Host Species: Mouse
Clone: PY20
Isotype: IgG2b
Species Reactivity: Many
Epitope: Phosphotyrosine
Immunogen: Phosphotyrosine conjugated to carrier protein
ELISA, Western Blotting

Specifications
Phosphotyrosine | |
PY20 | |
Mouse | |
10 Blots | |
Cell Signaling | |
Semi-purified |
Monoclonal | |
HRP | |
Phosphotyrosine conjugated to carrier protein | |
RUO | |
Primary | |
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.