Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-tyrosine Mouse anti-All, HRP, Clone: 4G10™ and PY20, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 16316
Description
Detect Phosphotyrosine with Anti-Phosphotyrosine Antibody, 4G10 Platinum, HRP Conjugate (Mouse Monoclonal Antibody), that has been shown to work in western blot, ELISA.
Specifications
Phosphotyrosine | |
Monoclonal | |
HRP | |
Varies | |
4G10™ used Phosphotyramine coupled to KLH and PY20 used phosphotyrosine conjugated to carrier protein | |
100 μL | |
All species | |
Purified |
ELISA, Western Blot | |
4G10™ and PY20 | |
100μL of a proprietary mixture of protein G purified mouse monoclonal 4G10™ (IgG2bƙ) HRP conjugate and protein A purified mouse monoclonal PY20, HRP conjugate (IgG2b) in PBS containing 0.05% ProClin™150 and HRP stabilizing compounds. Suffi | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction