Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphotyrosine Mouse, PE, Clone: PY20, BD

Catalog No. BDB558008 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558008 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558008 Supplier BD Biosciences Supplier No. 558008
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Phosphorylation of specific tyrosine residues is the result of activation or stimulation of their respective protein tyrosine kinases. The phosphorylated proteins can be autophosphorylated kinases or certain protein substrates that are regulated in oncogenesis or cell growth. Antibodies to phosphotyrosine provide one of the best tools for the detection and characterization of phosphotyrosine proteins. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: PY20
Isotype: IgG2b
Cross Reactivity: Human

Flow Cytometry, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphotyrosine
Applications Flow Cytometry
Classification Monoclonal
Clone PY20
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.