Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

putative homeodomain transcription factor 1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004510 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-510 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-510 Supplier Abnova Corporation Supplier No. H00010745A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PHTF1.

Sequence: REKPKFSDGEKCRREAFRRLGNGVSDDLSSEEDGEARTQMILLRRSVEGASSDNGCEVKNRKSILSRHLNSQVKKTTTRWCHIVRDSDSLAESEFESAAF

Specifications

Antigen putative homeodomain transcription factor 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PHTF1.
Formulation 50% glycerol
Gene PHTF1
Gene Accession No. NM_006608
Gene Alias PHTF
Gene Symbols PHTF1
Host Species Mouse
Immunogen PHTF1 (NP_006599, 248 a.a. ∼ 347 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10745
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.