Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI-16 Mouse anti-Human, PE, Clone: CRCBT-02-001, BD

Catalog No. BDB563520 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563520 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563520 Supplier BD Biosciences Supplier No. 563520

Mouse Monoclonal Antibody

The CRCBT-02-001 monoclonal antibody specifically binds to human CD364 which is also known as Peptidase inhibitor 16 (PI-16) or Protease inhibitor 16. PI-16 is a glycophosphatidylinositol-linked, 436 amino acid surface glycoprotein. It is a member of the cysteine-rich secretory protein (CRISP) family and is likewise known as CRISP9. PI-16 is a putative serine protease inhibitor and can serve as a binding protein for PSP94 (prostate secretory protein of 94 amino acids) termed PSPBP. Serum levels of PSP94 and PI-16 can reportedly serve as key targets for prostate cancer research. PI-16 is highly expressed by the activated/memory, FoxP3-bright subsets of regulatory T cells. PI-16-positive regulatory T cells expressed uniformly high levels of the chemokine receptors, CCR4 and CCR6, when compared with PI-16-negative T regulatory cells. The results suggest that the CCR4+CCR6+ regulatory T cells are capable of homing to inflammatory sites and regulating effector T cells such as CCR4+CCR6+ Th17-like cells. PI-16 is also expressed by subsets of memory CD4+ and CD8-bright T cells. It is not expressed by B cells or monocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PI-16
Applications Flow Cytometry
Classification Monoclonal
Clone WCHRI-1
Conjugate PE
Description PI16; Peptidase inhibitor 16; CRISP-9; PSPBP; PSP94-binding protein; MSMBBP
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human PI16 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.