Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI3 kinase alpha Mouse anti-Mouse, Clone: 2B12.1, MilliporeSigma™

Catalog No. MABS76MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS76MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS76MI Supplier MilliporeSigma Supplier No. MABS76

Mouse Monoclonal Antibody

This Anti-PI3 kinase alpha Antibody, clone 2B12.1 is validated for use in western blotting for the detection of PI3 kinase alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI3 kinase alpha
Applications Western Blot
Classification Monoclonal
Clone 2B12.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_852556
Gene Symbols PIK3R1, GRB1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human PI3 kinase alpha.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.