Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PI3 Kinase Mouse anti-Human, Mouse, Rat, Clone: 17D7.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051559
Description
Anti-PI3 Kinase Antibody, p110γ, clone 17D7.2 detects level of PI3 Kinase and has been published and validated for use in western blotting and immunocytochemistry.
Specifications
| PI3 Kinase | |
| Monoclonal | |
| Unconjugated | |
| NP_002640 | |
| Mouse | |
| Ion Exchange | |
| Primary | |
| 2-8°C for one year | |
| IgM κ |
| Immunocytochemistry, Western Blot | |
| 17D7.2 | |
| Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide. | |
| PI3CG, p120-PI3K, PIK3, PI3K, PIK3CG | |
| KLH-conjugated linear peptide corresponding to human PI3 Kinase, p110γ. | |
| 100 μg | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction