Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI3-Kinase Mouse, Unlabeled, Clone: 4, BD

Catalog No. BDB610046 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610046 150 μg
BDB610045 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610046 Supplier BD Biosciences Supplier No. 610046
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PI3-kinase phosphorylates the D-3 position of the inositol ring of phosphatidylinositol (PtdIns), PtdIns(4)P and PtdIns(4,5)P2 to produce the respective PI3-phosphorylated derivatives. PI3-kinase exists as a heterodimer of 85 kDa (p85) and 110 kDa (p110) subunits. The p85 subunit contains two SH2 domains and an SH3 domain. It associates with and serves as a substrate for activated growth factor receptor tyrosine kinases. p85 may serve as regulator of the catalytic subunit, p110, by acting as the link between PI3-kinase and the ligand-activated receptor. Two distinct forms of the p85 subunit have been described: 1) p85 α; , which binds tightly to the catalytic subunit, and 2) p85ß, a protein whose function is presently unknown. Both isoforms bind to activated receptors and serve as tyrosine kinase substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI3-Kinase
Applications Western Blot
Classification Monoclonal
Clone 4
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human PI3-Kinase α subunit aa. 562 to 724
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.