Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI3 Kinase Mouse anti-Human, Mouse, Rat, Clone: AB6, MilliporeSigma™

Catalog No. 05212MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-212-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-212-MI Supplier MilliporeSigma Supplier No. 05212
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-PI3 Kinase Antibody, p85, N-SH3, clone AB6 is validated for use in immunocytochemistry, immunoprecipitation, western blotting for the detection of PI3 Kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI3 Kinase
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AB6
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing PBS, pH 7.2 with 50% glycerol.
Gene Accession No. NM_181504
Gene Symbols PIK3R1, GRB1, PI3K, p85-ALPHA
Host Species Mouse
Immunogen Recombinant human p85 alpha PI3 Kinase expressed in E. Coli. Clone AB6.
Purification Method Protein A chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.