Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI3 Kinase p110δ Mouse, Unlabeled, Clone: 29, BD

Catalog No. BDB611014 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611014 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611014 Supplier BD Biosciences Supplier No. 611014

Mouse Monoclonal Antibody

Phosphoinositide 3 Kinase (PI3 Kinase) phosphorylates the D-3 position of the inositol ring of phosphatidylinositol, producing PtdIns(3)P, PtdIns(3,4)P2, and PtdIns (3,4,5)P3. PI3-kinase is a heterodimer of an 85kDa regulatory subunit (p85) containing two SH2 domains and an SH3 domain, and a 110kDa catalytic subunit (p110). Four isoforms of p110 have been identified (α , β , γ, and δ). The p110α, β isoforms are distributed ubiquitously. The p110δ isoform is found exclusively in lymphocytes and lymphoid tissues. Lymphocytes also contains p110α and both isoforms interact with p85 and are recruited to activated signaling complexes. However, several biochemical and structural differences distinguish p110δ. Unlike p110α, p110δ autophosphorylates and does not phosphorylate p85. In addition, p110δ contains a Pro-rich region, which endows it with the potential for interaction with SH3 domain-containing proteins, and a bZIP-like domain, which is a common transcription factor domain that has also been identified in certain protein kinases. With the established role of PI3-Ks in cytoskeletal rearrangements, it is thought that p110δ contributes specifically to the regulation of lymphocytes extravasation into tissues. There is a high degree of homology in the immunogen region with the P110β form (77%), and it is likely that our antibody will detect both the beta and delta forms.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PI3 Kinase p110δ
Applications Western Blot
Classification Monoclonal
Clone 29
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse PI3 Kinase p110δ aa. 73-90
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.