Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI3 Kinase p85α Mouse, Unlabeled, Clone: U15, BD

Catalog No. BDB556399 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556399 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556399 Supplier BD Biosciences Supplier No. 556399

Mouse Monoclonal Antibody

Phosphatidylinositol 3-kinase (PI3-kinase) is a universal enzyme associated with receptor signaling pathways. It generates second messenger phospholipids by phosphorylating the D-3 position of inositol phospholipids including phosphatidylinositol (PI), PI-4-phosphate, and PI-4, 5-biphosphate. The enzyme exists as a heterodimer composed of regulatory 85kDa (p85) and catalytic 110kDa (p110) subunits. The p85 subunit contains two SH2 (src-homology 2) domains and an SH3 domain. The catalytic activity of the p110 subunit is stimulated when the p85 regulatory subunit binds, through its SH2 domains, to activated receptor and non-receptor tyrosine kinases. Two p85 isoforms have been described, p85a and p85-». Both isoforms bind to activated receptors and each may be responsible for mediating a subset of PI3-kinase interactions. Clone U15 recognizes the p85 regulatory subunit of PI3 kinase (p85α). It reacts with human, monkey, and cow PI3 kinase. It does not cross-react with mouse or rat PI3 kinase. Recombinant cow p85α was used as immunogen. The epitope has been mapped to the inter-SH2 spacer region of p85α. The antibody will block lipids binding to this region.

Host Species: Mouse
Clone: U15
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant cow p85α

Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PI3 Kinase p85α
Applications Western Blot
Classification Monoclonal
Clone U15
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant cow p85α
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Bovine, Human, Monkey
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.