Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CALM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520871
Description
A suggested positive control is EL4 cell lysate. PA5-20871 can be used with blocking peptide PEP-0985.
PICALM, also known as the clathrin assembly lymphoid myeloid leukemia (CALM) protein, co-localizes with clathrin at clathrin-coated areas of the plasma membrane and in the Golgi area, and is thought to be an important component of the coated pit internalization machinery. Fusion proteins involving PICALM have been implicated in acute myeloid and T-cell acute lymphoblastic leukemia. PICALM is also expressed in neurons and is present at the synapse where it facilitates the endocytosis of the synaptic vesicle protein VAMP2. Recent studies have identified variants of PICALM associated with Alzheimer's disease and have suggested this protein may play a role in the endocytosis and recycling of the amyloid protein precursor (APP).
Specifications
CALM | |
Polyclonal | |
Unconjugated | |
Picalm | |
CALM; Clathrin assembly lymphoid myeloid leukemia; clathrin assembly lymphoid myeloid leukemia protein; clathrin-assembly lymphoid leukemia protein; clathrin-assembly lymphoid myeloid leukemia protein; CLTH; fit1; fit-1; LAP; mKIAA4114; PAP180; phosphatidylinositol binding clathrin assembly protein; phosphatidylinositol-binding clathrin assembly protein; PICALM; rCALM | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
233489, 8301, 89816 | |
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O55012, Q13492, Q7M6Y3 | |
Picalm | |
A 15 amino acid peptide from near the amino terminus of human PICALM. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction