Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piccolo Mouse anti-Mouse, Rat, Clone: 4G3.3, MilliporeSigma™

Catalog No. MAB2226MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2226MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2226MI Supplier MilliporeSigma Supplier No. MAB2226
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Pimmunocytochemistryolo Antibody, clone 4G3.3 is validated for use in western blot, immunohistochemistry (paraffin) for the detection of Pimmunocytochemistryolo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Piccolo
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4G3.3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS and 0.05% sodium azide.
Gene Accession No. NP_001104266
Gene Symbols ACZ , Aczonin, DKFZp779G1236, KIAA0559
Host Species Mouse
Immunogen GST-tagged recombinant protein
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.