Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pigment Epithelium Derived Factor Mouse anti-Human, Rat, Clone: 10F12.2, MilliporeSigma™

Catalog No. MAB1059MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1059MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1059MI Supplier MilliporeSigma Supplier No. MAB1059
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Pigment Epithelium Derived Factor Antibody, clone 10F12.2 is an antibody against Pigment Epithelium Derived Factor for use in ELISA, immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pigment Epithelium Derived Factor
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 10F12.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2a in buffer containing 0.02MPBS, pH 7.6, 0.25MNaCl with 0.1% sodium azide.
Gene Accession No. NM_002615.4
Gene Symbols SERPINF1, Serpin-F1, EPC-1, PEDF, PIG35
Host Species Mouse
Immunogen Recombinant human GST-PEDF fusion protein. The sequence corresponding to amino acids 196-347 of human PEDF tagged with GST on the N-terminal was expressed in E. Coli. GST-PEDF is about 46 kDa on SDS-PAGE.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.