Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIGN Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526755
Description
PIGN is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This protein is expressed in the endoplasmic reticulum and transfers phosphoethanolamine (EtNP) to the first mannose of the GPI anchor.
Specifications
| PIGN | |
| Polyclonal | |
| PIGN | |
| MCAHS, MCD4, MDC4, PIG-N, GPI ethanolamine phosphate transferase 1, MCD4 homolog, phosphatidylinositol-glycan biosynthesis class N protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23556, 27392 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| O95427, Q9R1S3 | |
| PIGN | |
| KLH conjugated synthetic peptide between 206-235 amino acids from the N-terminal region of human PIGN | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction