Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIK3CG Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA515506
Description
This antibody is predicted to react with mouse and porcine based on sequence homology.
PIK3CG belongs to the pi3/pi4-kinase family of proteins. It is an enzyme that phosphorylates phosphoinositides on the 3-hydroxyl group of the inositol ring. It is an important modulator of extracellular signals, including those elicited by E-cadherin-mediated cell-cell adhesion, which plays an important role in maintece of the structural and functional integrity of epithelia. In addition to its role in promoting assembly of adherens junctions, the protein is thought to play a pivotal role in the regulation of cytotoxicity in NK cells. The gene for this protein is located in a commonly deleted segment of chromosome 7 previously identified in myeloid leukemias.
Specifications
PIK3CG | |
Polyclonal | |
PIK3CG | |
Phosphatidylinositol-4;5-bisphosphate 3-kinase catalytic subunit gamma isoform; PI3-kinase p110 subunit gamma; PtdIns-3-kinase subunit p110; PI3K; PI3Kgamma; p120-PI3K; PIK3CG | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5294 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
P48736 | |
PIK3CG | |
KLH conjugated synthetic peptide between 1079-1102 amino acids from human PIK3CG | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction